Test strips, nitrate , Reflectoquant®, Supelco®
Nitrate test, 3 - 90 mg/l NO₃, Griess' reaction (modified), 50 Tests
: Merck
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
The Reflectoquant® Nitrate Test is designed for rapid, accurate of nitrate content in a variety of foods, such as vegetables or infant meals, determination and drinking water.
- Chemical method:Griess' reaction (modified)
- Pk:50 Tests
- Description:Nitrate test
- Range:3 - 90 mg/l NO₃
- Cat. No.:1.16995.0001
- M-Clarity quality level:MQ100