Order Entry
Sweden
ContactUsLinkComponent
FRACTOGEL EMD TMAE (M) STRONG ANION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (40-90 MYM) This product cannot be returned after delivery 1 * 500 ml
Catalog # 1.16881.0500
FRACTOGEL EMD TMAE (M) STRONG ANION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (40-90 MYM) This product cannot be returned after delivery 1 * 500 ml
Catalog # 1.16881.0500
Related Information