Celite® 545, filter aid, Supelco®
Plastic bottle, 1 kg
Leverantör: Merck
Gå till:
Product Details & Documents
Synonymer: DE, Diatomaceous earth, flux-calcined
- CAS-nummer:68855-54-9
- Formel:SiO₂
- MDL-nummer:MFCD00132803
- Molecular Weight:60.08 g/mol
- :M-Clarity™ quality level = MQ100
- EINECS:272-489-0
Documents
Certificates
Sök efter LOT-nummerMERC1.02693.0250_TDS_EN ProductPage_102693
Product specification
msds_102693
SDS
Specifikationer
Specifikationstestresultat
- Solubility:- 0.001 g/l (20 °C)
- Bulk Density:300 kg/m3
- pH:10 (100 g/l, H2O, 20 °C)
Specifikationer
- Pack type:Plastic bottle
- Pk:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



