Order Entry
Sweden
Orders LinkContactUsLinkComponent
Readycult®

Readycult®

Readycult® Coliforms 50*, 20 Tests
Catalog # 1.01295.0001
Supplier:   Merck
Readycult®
Catalog # 1.01295.0001
Supplier:  Merck
CAS Number:  
Readycult®
Readycult®

Readycult® Coliforms 50*, 20 Tests

CAS Number:  
Price
1 640,00 kr
Log in for availability
1 * 20 Tests
Product Details & Documents

Readycult® Coliform and Readycult® Enterococci make it easy to detect coliforms, E. coli or faecal streptococci in water. You only need a sterile container and a UV lamp (Cat. No. 1.13203.0001).

  • Easy handling: Readycult® is ready to use in a blister pack
  • Quick results: Add Readycult® to the water sample straight from the blister pack, mix, incubate for 24 ±1 hour, read result, finished!
  • Safe and easy interpretation: Readycult® reliably detects even damaged organisms. Blue-green colour change shows the presence of Total coliforms, E. coli or Enterococci (Cat. No. 1.01299); fluorescence + indole reaction shows the presence of E. coli. * for 50 ml water; ** for 100 ml water
Specifications

Specifications

  • Pk:
    20 Tests
  • Description:
    Readycult® Coliforms 50*

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.