Order Entry
Sweden
Orders LinkContactUsLinkComponent
Stopcocks, Nalgene®

Stopcocks, Nalgene®

2 mm, 6,35 - 9,53 mm
Catalog # 229-0016
   Low Manufacturing Impact
Stopcocks, Nalgene®
Catalog # 229-0016
   Low Manufacturing Impact
Stopcocks, Nalgene®
Stopcocks, Nalgene®

2 mm, 6,35 - 9,53 mm

Price
775,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Polypropylene with PTFE plug.

  • Autoclavable
  • Very good chemical resistance
  • Serrated tubulation on each end accepts 1/2" through 5/16" tubing

Permit reliable, accurate liquid flow control in liquid handling systems. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.

Specifications
  • Hole Ø:
    2 mm
  • For tube I-Ø:
    6,35 - 9,53 mm
  • Cat. No.:
    229-0016
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.