Order Entry
Sweden
Orders LinkContactUsLinkComponent
Pyrocatechol 99+%

Pyrocatechol 99+%

Glass bottle for solids, 500 g
Catalog # ACRO158985000
Corrosive
Toxic
Health
Pyrocatechol 99+%
Catalog # ACRO158985000
CAS Number:  120-80-9
Pyrocatechol 99+%
Pyrocatechol 99+%

Glass bottle for solids, 500 g

CAS Number:  120-80-9
Price
1 975,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,08089
  • Boiling Pt:245 °C (760 mmHg)
  • Melting Pt:103…106 °C
  • Density:1,0548 g/cm³ (20 °C)
  • Flash Pt:131 °C
  • Storage Temperature:Ambient
  • EINECS:204-427-5
  • UN:2811
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Off-white to grey-brown Crystalline flakes (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    103 to 106 °C
  • GC:
    ≥99.0 %
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.05 % (in water)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.