Thermometer drying oven straight stem, -10+100:1c, cap. blue reflecting, milk glass scale with metal clasp, mercury filled, upper p art c. 200x17mm, lower part 150x7,5mm, pin-top finish Until 10th October 2017 it is still allowed to use mercury the 1 * 1 items
Thermometer drying oven straight stem, -10+100:1c, cap. blue reflecting, milk glass scale with metal clasp, mercury filled, upper p art c. 200x17mm, lower part 150x7,5mm, pin-top finish Until 10th October 2017 it is still allowed to use mercury the 1 * 1 items