Order Entry
Sweden
ContactUsLinkComponent
Thermometer, Anschutz milk glass type, mercury filled, DIN 12777, measuring range:48:102/0.2C, enclosed scale, capillary lens fron t , colourless, dimension: 175x5-6mm, total immersion, Richter-top finish, suitable for government verification. Unti 1 * 1 items
Catalog # AMARN60162
Supplier:  Amarell
Thermometer, Anschutz milk glass type, mercury filled, DIN 12777, measuring range:48:102/0.2C, enclosed scale, capillary lens fron t , colourless, dimension: 175x5-6mm, total immersion, Richter-top finish, suitable for government verification. Unti 1 * 1 items
Catalog # AMARN60162
Supplier:  Amarell