Thermometer, Anschutz milk glass type, mercury filled, measuring range:-48:102/0.1C, enclosed scale, capillary lens front, colourl e ss, dimension: 340x5-6mm, Richter-top finish, suitable for government verification. Until 10th October 2017 it is s 1 * 1 items
Thermometer, Anschutz milk glass type, mercury filled, measuring range:-48:102/0.1C, enclosed scale, capillary lens front, colourl e ss, dimension: 340x5-6mm, Richter-top finish, suitable for government verification. Until 10th October 2017 it is s 1 * 1 items