Order Entry
Sweden
ContactUsLinkComponent
Thermometer, precision, mercury filled, blue reflecting, DIN certified, measuring range:-10:100/1C, enclosed scale, capllary lens f ront, colourless, Richter-top finish with button, dimension:305x8-8.5mm, suitable for government verification. Until 1 * 1 items
Catalog # 620-1050
Supplier:  Amarell
Thermometer, precision, mercury filled, blue reflecting, DIN certified, measuring range:-10:100/1C, enclosed scale, capllary lens f ront, colourless, Richter-top finish with button, dimension:305x8-8.5mm, suitable for government verification. Until 1 * 1 items
Catalog # 620-1050
Supplier:  Amarell