Order Entry
Sweden
ContactUsLinkComponent
Thermometer, precision, mercury filled, DIN 12775, measuring range:0:100/0.2C, enclosed scale, capllary lens front, colourless, Ri c hter-top finish with button, dimension:420x8.5-9mm, suitable for government verification. Until 10th October 2017 i 1 * 1 items
Catalog # 823-5948
Supplier:  Amarell
Thermometer, precision, mercury filled, DIN 12775, measuring range:0:100/0.2C, enclosed scale, capllary lens front, colourless, Ri c hter-top finish with button, dimension:420x8.5-9mm, suitable for government verification. Until 10th October 2017 i 1 * 1 items
Catalog # 823-5948
Supplier:  Amarell