Order Entry
Sweden
ContactUsLinkComponent
Thermometer, precision, mercury filled, DIN 12778, measuring range:0:360/1C, enclosed scale, capllary lens front, colourless, Rich t er-top finish with button, dimension:380x8-8.5mm, suitable for government verification. Until 10th October 2017 it 1 * 1 items
Catalog # AMARL25910
Supplier:  Amarell
Thermometer, precision, mercury filled, DIN 12778, measuring range:0:360/1C, enclosed scale, capllary lens front, colourless, Rich t er-top finish with button, dimension:380x8-8.5mm, suitable for government verification. Until 10th October 2017 it 1 * 1 items
Catalog # AMARL25910
Supplier:  Amarell