Order Entry
Sweden
ContactUsLinkComponent
Thermometer, mercury, ASTM39C, white backed, measuring range:48:102/0.2C, length:395mm, 100mm immersion, durable pigment, suitable for government calibration. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclu 1 * 1 items
Catalog # 620-0978
Supplier:  Amarell
Thermometer, mercury, ASTM39C, white backed, measuring range:48:102/0.2C, length:395mm, 100mm immersion, durable pigment, suitable for government calibration. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclu 1 * 1 items
Catalog # 620-0978
Supplier:  Amarell