Electroporation Cuvettes Plus
Cuvettes Plus with 4,0 mm gap size, yellow cap, 80 - 800 µl, Sterile
Supplier: BTX
Jump to:
Product Details & Documents
High quality disposable cuvettes for transfection available in three sizes and compatible with electroporation systems.
- Transfer pipette for manipulation of samples and gamma irradiated together for sterility
- Polished aluminium electrodes in the cuvettes lowers arcing frequency
- Cuvettes (without cap) are autoclavable
Cuvette caps are round for easy, one-handed removal, and are colour-coded for quick identification.
Documents
Specifications
- Sterility:Sterile
- Volume:80 - 800 µl
- Description:Cuvettes Plus with 4,0 mm gap size, yellow cap
- Seq:5
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cuvette rack with 20 numbered positions