Orange G, pure, certified
Glass bottle for solids, 10 g
Leverantör: Thermo Fisher Scientific
Orange G, pure, certified
Katalog # ACRO416550100
Leverantör: Thermo Fisher Scientific
CAS-nummer: 1936-15-8
Gå till:
Product Details & Documents
Synonymer: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10
- CAS-nummer:1936-15-8
- Formel:C₁₆H₁₀N₂Na₂O₇S₂
- MDL-nummer:MFCD00012457
- Molecular Weight:452.38 g/mol
- Smält Pt:141 °C
- Densitet:0,45 to 0.47 g/cm³ (20 °C)
- Förvaringstemperatur:Ambient
- EINECS:217-705-6
Documents
ACRO41655
SDS
Specifikationer
Specifikationstestresultat
- Appearance:Orange Powder
- Infrared spectrum:Conforms
- Dye content:≥80 %
- UV Lambda max:476 to 481 nm
- Absorption Ratio P-15/P+15:0.88 to 0.95
- BSC Certification:Passes
- Microbial contamination:≤10 /g (Escherichia coli)
Specifikationer
- Pack type:Glass bottle for solids
- Pk:10 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



