Order Entry
Sweden
Orders LinkContactUsLinkComponent
Cork borers
Cork borers
Cork borers (18), 5 − 26 mm
Catalog # 217-0706
Supplier:   BOCHEM
Cork borers
Catalog # 217-0706
Supplier:  BOCHEM
Cork borers
Cork borers
Cork borers (18), 5 − 26 mm
Price
5 325,00 kr
Log in for availability
1 * 1 SET
Product Details & Documents
Cork borers made of hard coated nickel-plated brass.
 : Include a cleaning needle.
Specifications
  • Ø:
    5 − 26 mm
  • Description:
    Cork borers (18)