Order Entry
Sweden
Orders LinkContactUsLinkComponent
Visual immunoassays
Staphylococcus enterotoxin, VIA kit, 96 well, 96
Catalog # 709-0255
Supplier:   TECRA
Visual immunoassays
Catalog # 709-0255
Supplier:  TECRA
CAS Number:  
Visual immunoassays
Visual immunoassays
Staphylococcus enterotoxin, VIA kit, 96 well, 96
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 96 Items
Product Details & Documents
Microtitre-format screening tests with high-affinity, specific antibodies for food and environment samples. Fast, specific bacteria and toxin testing with advanced Visual Immunoassay for food, water, environment samples, raw materials, cosmetics and pharmaceuticals. Ideal for rapid detection of pathogenic bacteria, 46 or 94 tests per plate.
  • The simplicity of the VIA™ method frees up time for other work and projects
  • Rapid results allow products to be released earlier, thereby saving on warehousing costs
  • The kit can be used manually, semi automatically or fully automatically for large numbers of samples
  • Fast results
  • High sensitivity and specificity
Specifications
Specifications
  • Pk:
    96
  • Description:
    Staphylococcus enterotoxin, VIA kit, 96 well
Specifications