Order Entry
Sweden
Orders LinkContactUsLinkComponent
Preparation forceps
Preparation forceps
Straight, fine, 105 mm
Catalog # HAMMHSC184-10
Supplier:   HAMMACHER KARL
Preparation forceps
Catalog # HAMMHSC184-10
Supplier:  HAMMACHER KARL
Preparation forceps
Preparation forceps
Straight, fine, 105 mm
Price
505,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Straight, fine
  • Length:
    105 mm
  • Seq:
    03