Order Entry
Sweden
Orders LinkContactUsLinkComponent
Preparation forceps
Preparation forceps
Straight, fine, 80 mm
Catalog # HAMMHSC180-08
Supplier:   HAMMACHER KARL
Preparation forceps
Catalog # HAMMHSC180-08
Supplier:  HAMMACHER KARL
Preparation forceps
Preparation forceps
Straight, fine, 80 mm
Price
550,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Straight, fine
  • Length:
    80 mm
  • Seq:
    01