0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Acro 50A syringe filter with HT tuffryn membrane 0.65/0.2Ám, 50mm, sterile 1 * 18 items
514-4112
:
Cytiva (Formerly Pall Lab)
Acro 50A syringe filter with HT tuffryn membrane 0.65/0.2Ám, 50mm, sterile 1 * 18 items
514-4112
:
Cytiva (Formerly Pall Lab)