COD standards
COD standard solution, 1300 mg/l, 500 mL
Catalog # 701-0046
Supplier: REAGECON
Jump to:
Product Details & Documents
Ideal for use as control standards to verify that correct analysis for COD has taken place.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Concentration:1300 mg/l
- Pk:500 mL
- Description:COD standard solution