Fumaric acid 99%
Bottle
Catalog # APOSOR1013408-500G
New Product
Supplier: Apollo Scientific
Fumaric acid 99%
Catalog # APOSOR1013408-500G
Supplier: Apollo Scientific
New Product
CAS Number: 110-17-8
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: trans-1,2-Ethylentricarboxylic acid
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116,07 g/mol
Specifications
Specifications
- Pack type:Bottle



