Order Entry
Sweden
Orders LinkContactUsLinkComponent
3'-Nitropropiophenone

3'-Nitropropiophenone

Bottle
Catalog # APOSOR11232-1G
Irritant
New Product
Supplier:   Apollo Scientific
3'-Nitropropiophenone
Catalog # APOSOR11232-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  17408-16-1
3'-Nitropropiophenone

Bottle

CAS Number:  17408-16-1
Price
770,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  m-Nitropropiophenone, 1-(3-Nitrophenyl)propan-1-one
  • CAS Number:17408-16-1
  • Formula:C₉H₉NO₃
  • MDL Number:MFCD00051516
  • Molecular Weight:179.18 g/mol
  • Melting Pt:99…102 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications