Order Entry
Sweden
Orders LinkContactUsLinkComponent
Actarit 98%

Actarit 98%

Bottle
Catalog # APOSOR911806-250MG
New Product
Supplier:   Apollo Scientific
Actarit 98%
Catalog # APOSOR911806-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  18699-02-0
Actarit 98%

Bottle

CAS Number:  18699-02-0
Price
260,00 kr
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid
  • CAS Number:18699-02-0
  • Formula:C₁₀H₁₁NO₃
  • MDL Number:MFCD00196165
  • Molecular Weight:193.20 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications