Temephos
Temephos
Catalog # EHERC17220000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COP(=S)(OC)Oc1ccc(Sc2ccc(OP(=S)(OC)OC)cc2)cc1 Inchi: InChI=1S/C16H20O6P2S3/c1-17-23(25,18-2)21-13-5-9-15(10-6-13)27-16-11-7-14(8-12-16)22-24(26,19-3)20-4/h5-12H,1-4H3 IUPAC: [4-(4-dimethoxyphosphinothioyloxyphenyl)sulfanylphenoxy]-dimethoxy-sulfanylidene-λ^{5}-phosphane
- Packing group: III
Specifications
Specifications
- Description:Temephos
- UN No.:2811
- Cat. No.:EHERC17220000
- Formula:C₁₆H₂₀O₆P₂S₃
- Storage conditions:–18 ℃
- Hazard class description:GHS06, GHS07, GHS09
- GHS signal:Danger
- Synonym(s):Phosphorothioic acid, Op,Op'-(thiodi-4,1-phenylene) Op,Op,Op',Op'-tetramethyl ester; Phosphorothioic acid, O,O-dimethyl ester, O,O-diester with 4,4'-thiodiphenol (7CI); Phosphorothioic acid, O,O'-(thiodi-4,1-phenylene) O,O,O',O'-tetramethyl ester (9CI); Phosphorothioic acid, O,O'-(thiodi-p-phenylene) O,O,O',O'-tetramethyl ester (8CI); AC 52160; Abate; Abate 4E; Abathion; Azai-SS; Biothion; Biothon; Difenphos; Difos; Diphos; Diphos (pesticide); EI 52160; ENT 27165; Ecopro 1707; Experimental Insecticide 52,160; Lavifos; Mosq; Mosquit TB 10; Mosquit TB 100; Nephis; Nephis 1G; O,O,O',O'-Tetramethyl O,O'-thiodi-p-phenylene phosphorothioate; Procida; Temecal; Temefos; Temephos
- Molecular weight:466,47
- CAS No.:3383-96-8
- Shipping conditions:RT



