Order Entry
Sweden
Orders LinkContactUsLinkComponent
Mepronil

Mepronil

Mepronil
Catalog # EHERC14890000
Supplier:   EHRENSTORFER
Mepronil
Catalog # EHERC14890000
Supplier:  EHRENSTORFER
CAS Number:  
Mepronil
Mepronil

Mepronil

CAS Number:  
Price
1 375,00 kr
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: CC(C)Oc1cccc(NC(=O)c2ccccc2C)c1 Inchi: InChI=1S/C17H19NO2/c1-12(2)20-15-9-6-8-14(11-15)18-17(19)16-10-5-4-7-13(16)3/h4-12H,1-3H3,(H,18,19) IUPAC: 2-methyl-N-(3-propan-2-yloxyphenyl)benzamide
  • Packing group: III
Specifications

Specifications

  • Description:
    Mepronil
  • UN No.:
    3077
  • Cat. No.:
    EHERC14890000
  • Formula:
    C₁₇H₁₉NO₂
  • Storage conditions:
    20 ℃
  • Hazard class description:
    GHS07, GHS09
  • GHS signal:
    Warning
  • Synonym(s):
    2-Methyl-N-[3-(1-methylethoxy)phenyl]benzamide; 2-Methyl-3'-isopropoxybenzanilide; 3'-Isopropoxy-2-methylbenzanilide; 3'-Isopropoxy-o-toluanilide; Basitac; Mepronil; Mepronil (pesticide)
  • Molecular weight:
    269,34
  • CAS No.:
    55814-41-0
  • Shipping conditions:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.