Metoxuron
Metoxuron
EHERC15180000
- SMILES: COc1ccc(NC(=O)N(C)C)cc1Cl
- Inchi: InChI=1S/C10H13ClN2O2/c1-13(2)10(14)12-7-4-5-9(15-3)8(11)6-7/h4-6H,1-3H3,(H,12,14)
- IUPAC: 3-(3-chloro-4-methoxyphenyl)-1,1-dimethylurea
- Packing group: III
- Description:Metoxuron
- UN No.:3077
- Cat. No.:EHERC15180000
- Formula:C₁₀H₁₃ClN₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Urea;3-(3-chloro-4-methoxyphenyl)-1,1-dimethyl- (6CI,8CI);N'-(3-Chloro-4-methoxyphenyl)-N,N-dimethylurea;1-(3-Chloro-4-methoxyphenyl)-3,3-dimethylurea;3-(3-Chloro-4-methoxyphenyl)-1,1-dimethylurea;Dosanex;Metoxuran;Metoxuron;N-(3-Chloro-4-methoxyphenyl)-N',N'-dimethylurea;Purivel;SAN 6602;SAN 7102;Urea;N'-(3-chloro-4-methoxyphenyl)-N,N-dimethyl-
- Molecular weight:228,68
- CAS No.:19937-59-8
- Shipping conditions:RT
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



