9-Methylanthracene
9-Methylanthracene
Catalog # EHERC20840000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cc1c2ccccc2cc3ccccc13
- Inchi: InChI=1S/C15H12/c1-11-14-8-4-2-6-12(14)10-13-7-3-5-9-15(11)13/h2-10H,1H3
- IUPAC: 9-methylanthracene
- Packing group: III
Specifications
Specifications
- Description:9-Methylanthracene
- UN No.:3077
- Cat. No.:EHERC20840000
- Formula:C₁₅H₁₂
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS07, GHS09
- GHS signal:Warning
- Molecular weight:192,26
- CAS No.:779-02-2
- Shipping conditions:RT



