Methyl violet 2B indicator
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
- CAS Number:8004-87-3
- Formula:C₂₄H₂₈ClN₃
- Molecular Weight:393,95 g/mol
5 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
- CAS Number:8004-87-3
- Formula:C₂₄H₂₈ClN₃
- Molecular Weight:393,95 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



