G-Biosciences Phosphate Buffered Saline, Sterile (1X and 10X)
Supplier: G-Biosciences
Certificates
About this item
Ready-to-use, sterile, pH adjusted blend of phosphate buffer and saline solution.
Sterile filtered through 0,2 µm filter
- 1X: 8 mM NaH₂PO₄, 150 mM NaCl, 3 mM KCl, 2 mM KH₂PO₄ (pH 7,4)
- 10X: 80 mM NaH₂PO₄ 1,5 M NaCl, 30 mM KCl, 20 mM KH₂PO₄
10X PBS (Phosphate Buffered Saline) is a 10 fold concentrated, sterile, pH adjusted blend of phosphate buffer and saline solution, which when diluted 1 to 10 in ultrapure water produces a phosphate buffered saline of 8 mM NaH₂PO₄, 150 mM NaCl, 3 mM KCl, and 2 mM KH₂PO₄ at pH 7,4. The 10X solution is a sterile solution.
1X PBS (Phosphate Buffered Saline) is a ready-to-use, sterile, pH adjusted blend of phosphate buffer and saline solution of 8 mM NaH₂PO₄, 150 mM NaCl, 3 mM KCl, and 2 mM KH₂PO₄ at pH 7,4.
Frequently Bought Together