Glass Fibre Extraction Thimbles, G400
Supplier: Sartorius
Certificates
About this item
These glass fibre thimbles are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives).
Composed of glass fibre cellulose for uncontaminated results
Engineered for superior mechanical stability to withstand rigorous use
Glass fibre thimbles offer remarkable resistance to high temperatures
Designed for a precise fit across a diverse range of extractors
Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
They are well suited to high temperature applications and are free of binders with high purity.