Order Entry
Sweden
ContactUsLinkComponent
NANOCOLOR® Photometric Standard Test Kit for Nickel
NANOCOLOR® Photometric Standard Test Kit for Nickel
Catalog # MANA91862
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Standard Test Kit for Nickel
Catalog # MANA91862
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Dimethylglyoxime
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Nickel, test no. 1-62
  • Dimensions:
    160×110×130 mm (including packaging)
  • Model:
    MANA91862
  • Platform:
    NANOCOLOR® standard tests
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    24
  • Weight:
    526.8 g (including packaging)
  • For unit:
    UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
  • Tests per kit:
    250
  • Range:
    0,01 - 10,0 mg/L (Ni²⁺)

Specifications

About this item

Photometric test kit for the determination of nickel in water samples.

  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II

Delivery: Rugged box with reagents for 250 tests.