Order Entry
Sweden
ContactUsLinkComponent
pan-HMR GSK3 beta ELISA Kit
pan-HMR GSK3 beta ELISA Kit
Catalog # ABCAAB315067-96TST
Supplier:  Abcam
New Product
CAS Number:  
undefined
pan-HMR GSK3 beta ELISA Kit
Catalog # ABCAAB315067-96TST
Supplier:  Abcam
New Product
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    GSK3B
  • Description:
    pan-HMR GSK3 beta ELISA kit
  • Sample type:
    Cell lysate
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    312,5 - 20000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    34,77 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

pan-HMR GSK3 beta ELISA kit is a single-wash 90-min Simplestep used to quantify pan-HMR GSK3 beta with a sensitivity of 34.77 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader .