HexArmor® Protective Sleeves, 60982 Series
Supplier: UVEX
Certificates
About this item
Industrial puncture protection for the forearm with spandex wrist insert, thumbhole, and snap fasteners.
- High cut and needle resistance
- Won’t fall down like knit sleeves
- Launderable
Two layers of SuperFabric® material protect against hazards through a configuration of tiny guard plates layered onto performance fabrics. Lightweight design is cooler than knit or leather alternatives.
Specifications
- Catalog No:
- UVEX6098201
- UVEX6098203
- UVEX6098202
- UVEX6098205
- UVEX6098204
- UVEX6098206
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN 388:2016
- EN 388:2016
- EN 388:2016
- EN 388:2016
- EN 388:2016
- EN 388:2016