Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG2b Mouse Monoclonal Antibody [clone: 2B 10A8] (Alexa Fluor® 488)
Anti-IgG2b Mouse Monoclonal Antibody [clone: 2B 10A8] (Alexa Fluor® 488)
Catalog # ABCAAB172334-250
Supplier:  Abcam
undefined
Anti-IgG2b Mouse Monoclonal Antibody [clone: 2B 10A8] (Alexa Fluor® 488)
Catalog # ABCAAB172334-250
Supplier:  Abcam

Specifications

  • Size:
    250 µg
  • Pack type:
    Vial

Specifications

About this item

Mouse monoclonal [2B 10A8] Anti-Rat IgG2b heavy chain (Alexa Fluor® 488).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Secondary
Antigen: IgG2b
Clonality: Monoclonal
Clone: 2B 10A8
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat