Slam Safety Spectacles
Supplier: Bollé Safety
New Product
Certificates
About this item
Modern design spectacles with extra-easy fitting and comfort.
- Panoramic visual field
- Anti-scratch, anti-fog, anti-static coatings
- Ultra enveloping
- Comfort temples
Weight: 23 g.
Certifications: EN 166, EN 170 (clear lens), EN 172 (smoke lens).
Delivery: Supplied with adjustable cord.
Frequently Bought Together