Order Entry
Sweden
ContactUsLinkComponent
 
Dihydrojasmone ≥98.0% (by GC)
  TCIAD1440-25ML
 :  TCI
 :  
undefined
Dihydrojasmone ≥98.0% (by GC)
  TCIAD1440-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :166.26 g/mol
 :84°C
 :96°C
 :214-434-5
 :1128-08-1
 :C₁₁H₁₈O
 :MFCD00036480