
4-(4-Nitrophenylazo)resorcinol
: TCI
:74-39-5
:
:C₁₂H₉N₃O₄
:MFCD00007310
:259.22 g/mol
:14,05695
:200-808-5
:GHS07
- Appearance:Orange to Brown to Dark red powder to crystal
- Absorbance:min. 0.700 (5 mg/L, 552.0 to 562.0 nm)
- Solubility in EtOH:almost transparency
- Drying loss:max. 5.0 %
- Sensitiveness:to pass test