Order Entry
Sweden
ContactUsLinkComponent
 
1-Octanol ≥99.5% (by GC) for determining partition coefficients
  TCIAO0212-500ML
 :  TCI
 :  
undefined
1-Octanol ≥99.5% (by GC) for determining partition coefficients
  TCIAO0212-500ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %

 

 

 :
 :130.23 g/mol
 :14,06751
 :194°C
 :-16°C
 :–16°C
 :0,8246 g/cm³ (20 °C)
 :83°C
 :203-917-6
 :GHS07
 :GHS08
 :Ambient
 :111-87-5
 :H₃C(CH₂)₆CH₂OH
 :MFCD00002988