Order Entry
Sweden
ContactUsLinkComponent
 
Trenbolone
Catalog # EHERCA17607700
Supplier:  EHRENSTORFER
CAS Number:  
Trenbolone
Catalog # EHERCA17607700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
CAS Number:10161-33-8
Formula:C₁₈H₂₂O₂
MDL Number:MFCD00214395