Organic reference standard, Dibenzothiophene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20727000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:132-65-0
- Description:Dibenzothiophene 10 µg/ml in acetonitrile
- Formula:C₁₂H₈S
- Description:Dibenzothiophene