Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2-Bromoethyl)pyrrole
1-(2-Bromoethyl)pyrrole ≥97.0% (by GC)
1-(2-Bromoethyl)pyrrole ≥97.0% (by GC)
:
TCI
1-(2-Bromoethyl)pyrrole ≥97.0% (by GC)
:
TCI
:
78358-86-8
:
:
C₆H₈BrN
:
MFCD00191322
:
GHS07
:
Refrigerator
Appearance:
Colorless to Light orange to Yellow clear liquid
Purity (GC):
min. 97.0 %