Order Entry
Sweden
ContactUsLinkComponent
 
9-Decyn-1-ol ≥94.0% (by GC)
 :  TCI
 
undefined
9-Decyn-1-ol ≥94.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 94.0 %