Order Entry
Sweden
ContactUsLinkComponent
 
 
3,3'-Diaminobenzidine tetrahydrochloride hydrate ≥98.0% (by HPLC, total nitrogen) for biochemical research
 :  TCI
 
undefined
 
3,3'-Diaminobenzidine tetrahydrochloride hydrate ≥98.0% (by HPLC, total nitrogen) for biochemical research
 :  TCI

 

  • Appearance:
    White to Amber to Dark purple powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    max. 13.0 %
  • Peroxidase detection test:
    to pass test