Indol-3-ylacetonitrile ≥98.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:771-51-7
:
Formel:C₁₀H₈N₂
MDL-nummer:MFCD00005628
:156.19 g/mol
Förvaringstemperatur:Refrigerator
EINECS:212-232-1
FN:3439
ADR:6.1,III
:GHS06
Specifikationstestresultat
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:34.0 to 38.0 deg-C
- Solubility in Methanol:almost transparency