Order Entry
Sweden
ContactUsLinkComponent
 
Irritant
N-Octyl-β-D-glucopyranoside ≥96.0% (by GC) for biochemical research
Catalog # TCIAO0355-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
N-Octyl-β-D-glucopyranoside ≥96.0% (by GC) for biochemical research
Catalog # TCIAO0355-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Specific rotation [a]20/D:
    -24.0 to -31.0 deg (C=1, Methanol)
  • Solubility in Water:
    almost transparency
  • Suitability for protein-analysis:
    to pass test

Specification Test Results

About this item

 :
 :292.37 g/mol
Merck Index:14,06767
Melting Pt:110 °C
EINECS:249-887-8
 :GHS07
Storage Temperature:Freezer
CAS Number:29836-26-8
Formula:C₁₄H₂₈O₆
MDL Number:MFCD00063288