Order Entry
Sweden
ContactUsLinkComponent
(±)-γ-Hexalactone ≥98.0% (by GC)
(±)-γ-Hexalactone ≥98.0% (by GC)
  TCIAH0514-25ML
 :  TCI
 :  
undefined
(±)-γ-Hexalactone ≥98.0% (by GC)
  TCIAH0514-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :114.14 g/mol
 :220°C
 :1,03 g/cm³ (20 °C)
 :98°C
 :211-778-8
 :695-06-7
 :C₆H₁₀O₂
 :MFCD00005401