
Ammonium sulphamate ≥98.5% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7773-06-0
:
Formula:H₆N₂O₃S
MDL Number:MFCD00011429
:114.13 g/mol
Merck Index:14,00554
Melting Pt:131.1 °C
Density:Min, 1 g/cm³ (20 °C)
EINECS:231-871-7
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (Diazo Method):min. 98.5 %
- Solubility in Water:almost transparency