3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:66304-01-6
:
Formula:C₇H₄O₃S₂
MDL Number:MFCD00132960
:200.24 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:102.0 to 106.0 deg-C