Organic reference standard, 2,2'-Binaphthyl 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20660000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:612-78-2
- Description:2,2'-Binaphthyl 10 µg/ml in acetonitrile
- Formula:C₂₀H₁₄
- Description:2,2'-Binaphthyl